Assay Details
| Gene Symbol: PWWP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558424.2 | XP_005558481.1 | 2 - 3 | 1616 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PWWP2A| Gene Symbol: PWWP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558424.2 | XP_005558481.1 | 2 - 3 | 1616 | 76 | |
Set Membership: |
|