Assay Details
| Gene Symbol: TRIM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319626.1 | NP_001306555.1 | 7 - 8 | 1168 | 88 | |
| XM_015448860.1 | XP_015304346.1 | 7 - 8 | 1168 | 88 | ||
| XM_015448861.1 | XP_015304347.1 | 7 - 8 | 1191 | 88 | ||
| XM_015448862.1 | XP_015304348.1 | 7 - 8 | 1187 | 88 | ||
| XM_015448863.1 | XP_015304349.1 | 7 - 8 | 1389 | 88 | ||
| XM_015448864.1 | XP_015304350.1 | 7 - 8 | 1174 | 88 | ||
| XM_015448865.1 | XP_015304351.1 | 7 - 8 | 1297 | 88 | ||
| XM_015448866.1 | XP_015304352.1 | 7 - 8 | 1165 | 88 | ||
| XM_015448868.1 | XP_015304354.1 | 7 - 8 | 1160 | 88 | ||
| GenBank mRNA | AB169159.1 | - | 7 - 8 | 1168 | 88 | |
More Information
Set Membership: |
|

TRIM39
Genomic Map