Assay Details
| Gene Symbol: LOC102140323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456804.1 | XP_015312290.1 | 17 - 18 | 2686 | 123 | |
| XM_015456815.1 | XP_015312301.1 | 17 - 18 | 2657 | 123 | ||
| XM_015456820.1 | XP_015312306.1 | 16 - 17 | 2554 | 123 | ||
| XM_015456822.1 | XP_015312308.1 | 16 - 17 | 2531 | 123 | ||
| XM_015456823.1 | XP_015312309.1 | 15 - 16 | 2393 | 123 | ||
| XM_015456824.1 | XP_015312310.1 | 15 - 16 | 2296 | 123 | ||
| XM_015456827.1 | XP_015312313.1 | 13 - 14 | 1874 | 123 | ||
More Information
Set Membership: |
|

LOC102140323
Genomic Map