Assay Details
| Gene Symbol: ZNF841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590173.2 | XP_005590230.1 | 4 | 2850 | 99 | |
| XM_005590175.2 | XP_005590232.1 | 5 | 2904 | 99 | ||
| XM_005590176.2 | XP_005590233.1 | 6 | 2991 | 99 | ||
| XM_005590177.2 | XP_005590234.1 | 2 | 2591 | 99 | ||
| XM_015441637.1 | XP_015297123.1 | 6 | 2994 | 99 | ||
| XM_015441639.1 | XP_015297125.1 | 4 | 2653 | 99 | ||
More Information
Set Membership: |
|

ZNF841
Genomic Map