Assay Details
| Gene Symbol: NPY2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556135.2 | XP_005556192.1 | 2 | 1571 | 110 | |
| XM_005556140.2 | XP_005556197.1 | 2 | 1592 | 110 | ||
More Information
Set Membership: |
|

NPY2R
Genomic Map