Assay Details
| Gene Symbol: NPY5R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556209.2 | XP_005556266.1 | 5 | 2075 | 126 | |
| XM_005556210.2 | XP_005556267.1 | 5 | 2071 | 126 | ||
| XM_005556211.2 | XP_005556268.1 | 4 | 1940 | 126 | ||
| XM_015450989.1 | XP_015306475.1 | 5 | 2072 | 126 | ||
| XM_015450990.1 | XP_015306476.1 | 5 | 1588 | 126 | ||
| XM_015450991.1 | XP_015306477.1 | 5 | 2068 | 126 | ||
| XM_015450992.1 | XP_015306478.1 | 4 | 1456 | 126 | ||
More Information
Set Membership: |
|

NPY5R
Genomic Map