Assay Details
| Gene Symbol: TICAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287328.1 | NP_001274257.1 | 1 | 1220 | 77 | |
| XM_005587588.2 | XP_005587645.1 | 2 | 1493 | 77 | ||
| GenBank mRNA | AB446489.1 | - | 1 | 1220 | 77 | |
More Information
Set Membership: |
|

TICAM1
Genomic Map