Assay Details
| Gene Symbol: UPF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594464.2 | XP_005594521.1 | 9 - 10 | 1123 | 87 | |
| XM_005594465.2 | XP_005594522.1 | 8 - 9 | 1082 | 87 | ||
More Information
Set Membership: |
|

UPF3B
Genomic Map