Assay Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283777.1 | NP_001270706.1 | 4 - 5 | 738 | 87 | |
| GenBank mRNA | AY445036.1 | - | 4 - 5 | 738 | 87 | |
More Information
Set Membership: |
|

CD2
Genomic Map