Assay Details
| Gene Symbol: EPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284561.1 | NP_001271490.1 | 2 - 3 | 378 | 77 | |
| GenBank mRNA | M18189.1 | - | 2 - 3 | 375 | 77 | |
More Information
Set Membership: |
|

EPO
Genomic Map