Assay Details
| Gene Symbol: TFPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573682.2 | XP_005573739.1 | 2 - 3 | 420 | 106 | |
| XM_005573683.2 | XP_005573740.1 | 2 - 3 | 412 | 106 | ||
| XM_015432466.1 | XP_015287952.1 | 3 - 4 | 514 | 106 | ||
| XM_015432467.1 | XP_015287953.1 | 3 - 4 | 534 | 106 | ||
| XM_015432468.1 | XP_015287954.1 | 3 - 4 | 518 | 106 | ||
| GenBank mRNA | AB220483.1 | - | 2 - 3 | 384 | 106 | |
More Information
Set Membership: |
|

TFPI
Genomic Map