Assay Details
| Gene Symbol: PAFAH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582483.1 | XP_005582540.1 | 9 - 10 | 1572 | 95 | |
| GenBank mRNA | AB083321.2 | - | 9 - 10 | 1223 | 95 | |
| AB173091.1 | - | 9 - 10 | 1608 | 95 | ||
More Information
Set Membership: |
|

PAFAH1B1
Genomic Map