Assay Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552165.2 | XP_005552222.1 | 4 - 5 | 1322 | 113 | |
| XM_005552166.2 | XP_005552223.1 | 4 - 5 | 3055 | 113 | ||
More Information
Set Membership: |
|

FBXO5
Genomic Map