Assay Details
| Gene Symbol: FATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594838.2 | XP_005594895.1 | 1 - 2 | 217 | 61 | |
| XM_005594839.2 | XP_005594896.1 | 1 - 2 | 219 | 61 | ||
More Information
Set Membership: |
|

FATE1
Genomic Map