Assay Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594284.2 | XP_005594341.1 | 3 - 4 | 1066 | 77 | |
| XM_005594285.2 | XP_005594342.1 | 3 - 4 | 889 | 77 | ||
More Information
Set Membership: |
|

RNF128
Genomic Map