Assay Details
| Gene Symbol: FAM3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547502.2 | XP_005547559.1 | 8 - 9 | 771 | 75 | |
| XM_005547503.2 | XP_005547560.1 | 8 - 9 | 4041 | 75 | ||
| XM_015446125.1 | XP_015301611.1 | 8 - 9 | 962 | 75 | ||
| XM_015446126.1 | XP_015301612.1 | 8 - 9 | 782 | 75 | ||
More Information
Set Membership: |
|

FAM3D
Genomic Map