Assay Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567263.2 | XP_005567320.1 | 2 - 3 | 338 | 64 | |
| XM_005567264.2 | XP_005567321.1 | 2 - 3 | 305 | 64 | ||
More Information
Set Membership: |
|

DNAL4
Genomic Map