Assay Details
| Gene Symbol: MTERF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550258.2 | XP_005550315.1 | 1 - 2 | 58 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005550258.2 transcripts.
Set Membership: |
|

MTERF1
Genomic Map