Assay Details
| Gene Symbol: MTERF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550258.2 | XP_005550315.1 | 2 - 3 | 126 | 68 | |
| XM_005550259.2 | XP_005550316.1 | 2 - 3 | 134 | 68 | ||
More Information
Set Membership: |
|

MTERF1
Genomic Map