Assay Details
| Gene Symbol: RELCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586527.2 | XP_005586584.1 | 10 - 11 | 1874 | 101 | |
| XM_015439667.1 | XP_015295153.1 | 10 - 11 | 1874 | 101 | ||
| XM_015439668.1 | XP_015295154.1 | 10 - 11 | 1874 | 101 | ||
| XM_015439671.1 | XP_015295157.1 | 9 - 10 | 1802 | 101 | ||
More Information
Set Membership: |
|

RELCH
Genomic Map