Assay Details
| Gene Symbol: SSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550974.2 | XP_005551031.1 | 4 - 5 | 407 | 103 | |
| XM_005550975.2 | XP_005551032.1 | 4 - 5 | 312 | 103 | ||
| XM_005550976.2 | XP_005551033.1 | 4 - 5 | 445 | 103 | ||
| XM_005550977.2 | XP_005551034.1 | 4 - 5 | 349 | 103 | ||
| XM_005550979.2 | XP_005551036.1 | 4 - 5 | 294 | 103 | ||
| XM_005550980.2 | XP_005551037.1 | 4 - 5 | 407 | 103 | ||
| XM_005550981.1 | XP_005551038.1 | 4 - 5 | 516 | 103 | ||
| XM_005550982.1 | XP_005551039.1 | 4 - 5 | 357 | 103 | ||
| XM_005550984.1 | XP_005551041.1 | 4 - 5 | 534 | 103 | ||
More Information
Set Membership: |
|

SSBP1
Genomic Map