Assay Details
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539826.2 | XP_005539883.1 | 2 - 3 | 698 | 59 | |
| GenBank mRNA | AB169719.1 | - | 1 - 2 | 173 | 59 | |
| AB173368.1 | - | 1 - 2 | 173 | 59 | ||
More Information
Set Membership: |
|

RGS4
Genomic Map