Assay Details
| Gene Symbol: MGST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539847.2 | XP_005539904.1 | 3 - 4 | 624 | 105 | |
| XM_015444838.1 | XP_015300324.1 | 3 - 4 | 287 | 105 | ||
| XM_015444840.1 | XP_015300326.1 | 3 - 4 | 220 | 105 | ||
| GenBank mRNA | AB170781.1 | - | 3 - 4 | 234 | 105 | |
| AB174024.1 | - | 3 - 4 | 230 | 105 | ||
| KC734515.1 | - | 2 - 3 | 192 | 105 | ||
More Information
Set Membership: |
|

MGST3
Genomic Map