Assay Details
| Gene Symbol: OTUD6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563677.2 | XP_005563734.1 | 3 - 4 | 504 | 78 | |
| XM_005563678.2 | XP_005563735.1 | 3 - 4 | 504 | 78 | ||
More Information
Set Membership: |
|

OTUD6B
Genomic Map