Assay Details
Gene Symbol: NCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549408.2 | XP_005549465.1 | 5 - 6 | 551 | 63 | |
XM_005549409.2 | XP_005549466.1 | 5 - 6 | 551 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
