Assay Details
| Gene Symbol: TTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558426.2 | XP_005558483.1 | 3 - 4 | 511 | 79 | |
| GenBank mRNA | AB170206.1 | - | 3 - 4 | 497 | 79 | |
More Information
Set Membership: |
|

TTC1
Genomic Map