Assay Details
Gene Symbol: LOC102145630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569455.2 | XP_005569512.1 | 2 - 3 | 2745 | 104 | |
XM_005569460.2 | XP_005569517.1 | 2 - 3 | 167 | 104 | ||
XM_015430002.1 | XP_015285488.1 | 2 - 3 | 324 | 104 | ||
XM_015430013.1 | XP_015285499.1 | 2 - 3 | 257 | 104 | ||
XM_015430014.1 | XP_015285500.1 | 2 - 3 | 209 | 104 | ||
XM_015430015.1 | XP_015285501.1 | 2 - 3 | 494 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015430002.1, XM_015430013.1, XM_015430014.1, XM_015430015.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
