Assay Details
| Gene Symbol: LACTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563513.2 | XP_005563570.1 | 1 - 2 | 363 | 103 | |
| GenBank mRNA | AB170438.1 | - | 1 - 2 | 215 | 103 | |
More Information
Set Membership: |
|

LACTB2
Genomic Map