Assay Details
| Gene Symbol: TMEM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590390.2 | XP_005590447.1 | 5 - 6 | 788 | 65 | |
| XM_005590393.2 | XP_005590450.2 | 5 - 6 | 1101 | 65 | ||
| XM_015441877.1 | XP_015297363.1 | 5 - 6 | 1111 | 65 | ||
| XM_015441878.1 | XP_015297364.1 | 5 - 6 | 1104 | 65 | ||
| XM_015441880.1 | XP_015297366.1 | 5 - 6 | 790 | 65 | ||
| XM_015441881.1 | XP_015297367.1 | 5 - 6 | 1158 | 65 | ||
| XM_015441882.1 | XP_015297368.1 | 5 - 6 | 742 | 65 | ||
More Information
Set Membership: |
|

TMEM150B
Genomic Map