Assay Details
| Gene Symbol: NAXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586234.2 | XP_005586291.1 | 4 - 5 | 473 | 65 | |
| XM_005586235.2 | XP_005586292.1 | 4 - 5 | 515 | 65 | ||
| XM_015439521.1 | XP_015295007.1 | 3 - 4 | 1408 | 65 | ||
More Information
Set Membership: |
|

NAXD
Genomic Map