Assay Details
| Gene Symbol: ALG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585676.2 | XP_005585733.1 | 3 - 4 | 347 | 93 | |
| XM_005585678.2 | XP_005585735.1 | 3 - 4 | 467 | 93 | ||
More Information
Set Membership: |
|

ALG5
Genomic Map