Assay Details
| Gene Symbol: UBE2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283170.1 | NP_001270099.1 | 2 - 3 | 131 | 92 | |
| XM_005545648.2 | XP_005545705.1 | 2 - 3 | 427 | 92 | ||
| XM_005545650.2 | XP_005545707.1 | 2 - 3 | 1329 | 92 | ||
| GenBank mRNA | AB178995.1 | - | 2 - 3 | 131 | 92 | |
More Information
Set Membership: |
|

UBE2E1
Genomic Map