Assay Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576109.2 | XP_005576166.1 | 2 - 3 | 448 | 64 | |
| XM_005576110.2 | XP_005576167.1 | 2 - 3 | 481 | 64 | ||
| XM_005576111.2 | XP_005576168.1 | 2 - 3 | 1675 | 64 | ||
More Information
Set Membership: |
|

QPCT
Genomic Map