Assay Details
| Gene Symbol: DOHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587502.2 | XP_005587559.1 | 3 - 4 | 842 | 84 | |
| XM_005587503.2 | XP_005587560.1 | 2 - 3 | 411 | 84 | ||
| XM_005587504.2 | XP_005587561.1 | 2 - 3 | 900 | 84 | ||
| XM_005587505.2 | XP_005587562.1 | 2 - 3 | 1126 | 84 | ||
| XM_015440306.1 | XP_015295792.1 | 2 - 3 | 1088 | 84 | ||
More Information
Set Membership: |
|

DOHH
Genomic Map