Assay Details
| Gene Symbol: UBC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572601.2 | XP_005572658.1 | 1 - 2 | 651 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005572601.2 transcripts.
Set Membership: |
|

UBC
Genomic Map