Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565430.2 | XP_005565487.1 | 5 - 6 | 982 | 68 | |
| XM_005565432.2 | XP_005565489.1 | 5 - 6 | 688 | 68 | ||
| XM_005565433.2 | XP_005565490.1 | 5 - 6 | 1080 | 68 | ||
| XM_005565434.2 | XP_005565491.1 | 5 - 6 | 762 | 68 | ||
| XM_005565435.2 | XP_005565492.1 | 5 - 6 | 981 | 68 | ||
| GenBank mRNA | AB173936.1 | - | 4 - 5 | 577 | 68 | |
More Information
Set Membership: |
|

AP3M1
Genomic Map