Assay Details
| Gene Symbol: MRPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584996.2 | XP_005585053.1 | 6 - 7 | 1262 | 62 | |
| XM_005584997.2 | XP_005585054.1 | 6 - 7 | 1418 | 62 | ||
| XM_005584998.2 | XP_005585055.1 | 7 - 8 | 1505 | 62 | ||
More Information
Set Membership: |
|

MRPL38
Genomic Map