Assay Details
| Gene Symbol: ACAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283595.1 | NP_001270524.1 | 6 - 7 | 614 | 92 | |
| GenBank mRNA | AB083303.2 | - | 6 - 7 | 614 | 92 | |
More Information
Set Membership: |
|

ACAT1
Genomic Map