Assay Details
| Gene Symbol: AP2M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287724.1 | NP_001274653.1 | 7 - 8 | 892 | 64 | |
| XM_005546525.2 | XP_005546582.1 | 6 - 7 | 1015 | 64 | ||
| GenBank mRNA | AB169024.1 | - | 7 - 8 | 892 | 64 | |
| AB170348.1 | - | 7 - 8 | 810 | 64 | ||
More Information
Set Membership: |
|

AP2M1
Genomic Map