Assay Details
| Gene Symbol: IGSF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441936.1 | XP_015297422.1 | 3 - 4 | 2046 | 74 | |
| XM_015441937.1 | XP_015297423.1 | 4 - 5 | 1915 | 74 | ||
| XM_015441941.1 | XP_015297427.1 | 3 - 4 | 1356 | 74 | ||
More Information
Set Membership: |
|

IGSF23
Genomic Map