Assay Details
| Gene Symbol: EBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593461.1 | XP_005593518.1 | 4 - 5 | 777 | 62 | |
| XM_005593462.2 | XP_005593519.1 | 5 - 6 | 1324 | 62 | ||
More Information
Set Membership: |
|

EBP
Genomic Map