Assay Details
| Gene Symbol: MMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579468.2 | XP_005579525.1 | 5 - 6 | 921 | 134 | |
| XM_015435642.1 | XP_015291128.1 | 5 - 6 | 809 | 134 | ||
More Information
Set Membership: |
|

MMP1
Genomic Map