Assay Details
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283417.1 | NP_001270346.1 | 3 - 4 | 254 | 94 | |
GenBank mRNA | AB170275.1 | - | 3 - 4 | 252 | 94 | |
AB170889.1 | - | 3 - 4 | 254 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
