Assay Details
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283417.1 | NP_001270346.1 | 4 - 5 | 406 | 106 | |
GenBank mRNA | AB170275.1 | - | 4 - 5 | 404 | 106 | |
AB170889.1 | - | 4 - 5 | 406 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
