Assay Details
| Gene Symbol: TCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567585.2 | XP_005567642.1 | 4 - 5 | 892 | 91 | |
| XM_015457509.1 | XP_015312995.1 | 4 - 5 | 728 | 91 | ||
| XM_015457510.1 | XP_015312996.1 | 4 - 5 | 725 | 91 | ||
More Information
Set Membership: |
|

TCN2
Genomic Map