Assay Details
| Gene Symbol: SFT2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551458.2 | XP_005551515.1 | 7 - 8 | 561 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SFT2D1| Gene Symbol: SFT2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551458.2 | XP_005551515.1 | 7 - 8 | 561 | 100 | |
Set Membership: |
|