Assay Details
| Gene Symbol: TMEM106A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584330.2 | XP_005584387.1 | 3 - 4 | 442 | 74 | |
| XM_005584331.2 | XP_005584388.1 | 3 - 4 | 484 | 74 | ||
| XM_005584333.2 | XP_005584390.1 | 3 - 4 | 442 | 74 | ||
| XM_015438540.1 | XP_015294026.1 | 3 - 4 | 462 | 74 | ||
More Information
Set Membership: |
|

TMEM106A
Genomic Map