Assay Details
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577063.2 | XP_005577120.1 | 1 - 2 | 246 | 55 | |
| XM_015434372.1 | XP_015289858.1 | 1 - 2 | 247 | 55 | ||
More Information
Set Membership: |
|

AIP
Genomic Map