Assay Details
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567117.2 | XP_005567174.1 | 1 - 2 | 337 | 91 | |
| GenBank mRNA | AB173331.1 | - | 1 - 2 | 226 | 91 | |
More Information
Set Membership: |
|

SMDT1
Genomic Map