Assay Details
| Gene Symbol: KRBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546803.2 | XP_005546860.2 | 4 - 5 | 442 | 140 | |
| XM_005546804.2 | XP_005546861.2 | 4 - 5 | 438 | 140 | ||
| XM_015445722.1 | XP_015301208.1 | 4 - 5 | 508 | 140 | ||
| XM_015445723.1 | XP_015301209.1 | 4 - 5 | 348 | 140 | ||
More Information
Set Membership: |
|

KRBOX1
Genomic Map